Learn More
Cayman Chemical Ginkgolide B, Purity: ≥98%, CAS: 15291-77-7, 50 mg
SDP
Catalog No. 502972269
Encompass_Preferred
A potent PAFR antagonist that inhibits platelet aggregation at concentrations from 10-500 μM; demonstrates anticancer, anti-inflammatory, anti-allergic, antioxidant, and neuroprotective effects
Catalog No. 50-297-2269 Supplier Cayman Chemical Supplier No. 1463650 MG
A potent PAFR antagonist that inhibits platelet aggregation at concentrations from 10-500 μM; demonstrates anticancer, anti-inflammatory, anti-allergic, antioxidant, and neuroprotective effects