Learn More
Selleck Chemical LLC Ginkgolide B 25mg 15291-77-7 BN52021
SDP
Supplier:  Selleck Chemical LLC S134325MG
Encompass_Preferred
Ginkgolide B (BN52021) is a PAFR antagonist with IC50 of 3.6 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-730-3