Learn More
AdipoGen Ginkgolide C (50 mg)
SDP
Supplier:  AdipoGen CDXG0214M050
Encompass_Preferred
Chemical. CAS: 15291-76-6. Formula: C20H24O11. MW: 440.4. Isolated from plant source. Ginkgolide C is a flavone first extracted from G. biloba leaves with multiple reported biological functions. It has been identified as a selective platelet activating factor receptor antagonist. Ginkgolide C has also been shown to increase lipolysis and to inhibit adipogenesis in adipocytes via the activated AMPK pathway. Shown to inhibit CD40-NF-kappaB pathway. Can be used as a reference compound.
Catalog No. 50-205-4475