Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginkgolide C, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013218 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3218 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3218 Supplier MilliporeSigma Supelco Supplier No. 1830910MGF
Add to Cart
Add to Cart
Ginkgolide C is an active constituent of Ginkgo biloba. The ginkgo leaves has several pharmacological properties, some of them being; improved blood flow, radical scavenging, vasoprotection, and anti-PAF (platelet aggregating factor) activity. Ginkgolide C along with the other active constituents of Ginkgo biloba, is responsible for the inhibition of PAF activity. Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 15291-76-6
Physical Form Neat
Linear Formula C20H24O11
MDL Number MFCD02094178
Synonym 1,7-Dihydroxy-ginkgolide A
Type Analytical Standard
Quantity 10 mg
Formula Weight 440.40
Percent Purity ≥96% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.