Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ginkgolide K | 153355-70-5 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYN41761MG

Encompass_Preferred

Ginkgolide K, isolated from Ginkgo biloba, induces protective autophagy through the AMPK/mTOR/ULK1 signaling pathway. It also possesses neuroprotective activity.

  • Purity of 98.59%
  • Molecular weight of 406.38
  • Chemical formula C20H22O9
  • Appears as a solid
  • White to off-white color
  • Classified as terpenoids diterpenoids
  • Isolated from Ginkgo biloba
  • Soluble in DMSO at 50 mg/mL

Catalog No. 50-004-20408


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.