Learn More
Medchemexpress LLC Ginkgolide K | 153355-70-5 | 98.6% | 406.38 g/mol | C20H22O9 | 10 MG
SDP
Catalog No. 5000289518
Encompass_Preferred
Ginkgolide K is a purified natural product isolated from Ginkgo biloba that acts as an autophagy activator and has reported neuroprotective activity. Supplied as a white to off-white solid for research use, the compound includes solubility and storage guidance for in vitro and in vivo formulation.
  • Activates autophagy through the AMPK/mTOR/ULK1 signaling pathway.
  • Demonstrates reported neuroprotective and platelet antagonist activities.
  • High purity (≈98.6%) suitable for research applications.
  • Soluble in DMSO (50 mg/mL with ultrasonic assistance) with in vivo formulation protocols available.
  • Solid storage recommended at 4°C; in-solution stability guidance provided for -80°C and -20°C.
  • Available in small research pack sizes for assay development and preclinical studies.
Catalog No. 50-002-89518 Supplier Medchemexpress LLC Supplier No. HYN417610MG
Ginkgolide K is a purified natural product isolated from Ginkgo biloba that acts as an autophagy activator and has reported neuroprotective activity. Supplied as a white to off-white solid for research use, the compound includes solubility and storage guidance for in vitro and in vivo formulation.
  • Activates autophagy through the AMPK/mTOR/ULK1 signaling pathway.
  • Demonstrates reported neuroprotective and platelet antagonist activities.
  • High purity (≈98.6%) suitable for research applications.
  • Soluble in DMSO (50 mg/mL with ultrasonic assistance) with in vivo formulation protocols available.
  • Solid storage recommended at 4°C; in-solution stability guidance provided for -80°C and -20°C.
  • Available in small research pack sizes for assay development and preclinical studies.