Learn More
TARGETMOL CHEMICALS INC GINKGOLIDE K 20MG
SDP
Catalog No. 502579094
Encompass_Preferred
Also available in 5 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ginkgolide K induces protective autophagy through the AMPK/mTOR/ULK1 signalling pathway. It possesses neuroprotective activity. purity: 99%
Catalog No. 50-257-9094 Supplier TARGETMOL CHEMICALS INC Supplier No. T820020MG
Also available in 5 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ginkgolide K induces protective autophagy through the AMPK/mTOR/ULK1 signalling pathway. It possesses neuroprotective activity. purity: 99%