Learn More
Medchemexpress LLC HY-N4176 5mg Medchemexpress, Ginkgolide K CAS:153355-70-5 Purity:>98%
SDP
Catalog No. 501970430
Encompass_Preferred
Medchemexpress, HY-N4176 5mg Ginkgolide K CAS:153355-70-5 Ginkgolide K, isolated from Ginkgo biloba, induces protective autophagy through the AMPK/mTOR/ULK1 signaling pathway. Ginkgolide K possesses neuroprotective activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0430 Supplier Medchemexpress LLC Supplier No. HYN41765MG
Medchemexpress, HY-N4176 5mg Ginkgolide K CAS:153355-70-5 Ginkgolide K, isolated from Ginkgo biloba, induces protective autophagy through the AMPK/mTOR/ULK1 signaling pathway. Ginkgolide K possesses neuroprotective activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.