Learn More
Cayman Chemical GInkgonlIc AcId 10mg
SDP
Catalog No. 502940274
Encompass_Preferred
An anacardic acid with diverse biological activities; inhibits GPDH (IC50 = 3 μg/ml); inhibits PI3Kδ (IC50 = 2.49 μM) and IgE-mediated RBL-2H3 mast cell degranulation in vitro (IC50 = 2.4 μM); reduces S. mutans biofilm formation (MBIC50 = 4 μg/ml)
Catalog No. 50-294-0274 Supplier Cayman Chemical Supplier No. 2765610 MG
An anacardic acid with diverse biological activities; inhibits GPDH (IC50 = 3 μg/ml); inhibits PI3Kδ (IC50 = 2.49 μM) and IgE-mediated RBL-2H3 mast cell degranulation in vitro (IC50 = 2.4 μM); reduces S. mutans biofilm formation (MBIC50 = 4 μg/ml)