Learn More
Cayman Chemical GInkgotoxIn 10mg
SDP
Catalog No. 502950020
Encompass_Preferred
A pyridoxine antimetabolite; inhibits pyridoxal kinase (Ki = 3 µM); induces seizure-like behavior in zebrafish larvae at 200-1,000 µM
Catalog No. 50-295-0020 Supplier Cayman Chemical Supplier No. 3346010 MG
A pyridoxine antimetabolite; inhibits pyridoxal kinase (Ki = 3 µM); induces seizure-like behavior in zebrafish larvae at 200-1,000 µM