Learn More
Cayman Chemical Ginkgotoxin, Purity: ≥98%, CAS: 1464-33-1, 50 mg
SDP
Catalog No. 502946306
Encompass_Preferred
A pyridoxine antimetabolite; inhibits pyridoxal kinase (Ki = 3 µM); induces seizure-like behavior in zebrafish larvae at 200-1,000 µM
Catalog No. 50-294-6306 Supplier Cayman Chemical Supplier No. 3346050 MG
A pyridoxine antimetabolite; inhibits pyridoxal kinase (Ki = 3 µM); induces seizure-like behavior in zebrafish larvae at 200-1,000 µM