Learn More
Aviva Systems Biology Corp GINS1 Antibody - N-terminal region (ARP46298_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46298_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GINS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GINS1. Alternate names: KIAA0186, PSF1, RP4-691N24.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53468