Learn More
Cayman Chemical Ginsenoside F1, Purity: ≥98%, CAS: 53963-43-2, 10 mg
SDP
Catalog No. 502989282
Encompass_Preferred
A ginsenoside with diverse biological activities; enhances NK cytotoxicity against K562 cells in vitro at 10 µM and in vivo; reduces atherosclerotic lesion area, whole body MPO levels, and aortic root NF-kB, TLR4, and LOX-1 levels in ApoE-/- mice at 50 mg/kg; increases microvessel density and improves focal cerebral blood perfusion in a rat model of MCAO-induced ischemic stroke; restores spatial working memory in the APP/PS1 transgenic mouse model of Alzheimer's disease
Catalog No. 50-298-9282 Supplier Cayman Chemical Supplier No. 3113710 MG
A ginsenoside with diverse biological activities; enhances NK cytotoxicity against K562 cells in vitro at 10 µM and in vivo; reduces atherosclerotic lesion area, whole body MPO levels, and aortic root NF-kB, TLR4, and LOX-1 levels in ApoE-/- mice at 50 mg/kg; increases microvessel density and improves focal cerebral blood perfusion in a rat model of MCAO-induced ischemic stroke; restores spatial working memory in the APP/PS1 transgenic mouse model of Alzheimer's disease