Learn More
TARGETMOL CHEMICALS INC GINSENOSIDE F1 25MG
SDP
Catalog No. 502580283
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ginsenoside F1 (20(S)-Ginsenoside F1) is an enzymatically modified derivative of ginsenoside Rg1 showing competitive inhibition of the activity of CYP3A4 and a weaker inhibition of the activity of CYP2D6. purity: 99%
Catalog No. 50-258-0283 Supplier TARGETMOL CHEMICALS INC Supplier No. T340125MG
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ginsenoside F1 (20(S)-Ginsenoside F1) is an enzymatically modified derivative of ginsenoside Rg1 showing competitive inhibition of the activity of CYP3A4 and a weaker inhibition of the activity of CYP2D6. purity: 99%