Learn More
Apexbio Technology LLC Ginsenoside F2 62025-49-4 20mg
SDP
Supplier:  Apexbio Technology LLC N156820
Encompass_Preferred
Ginsenoside F2 (CAS 62025-49-4) is a saponin compound isolated from Panax ginseng C A Mey It has been studied for its modulatory effects on cellular signaling pathways involved in cell proliferation apoptosis and metabolic regulation Ginsenoside F2 is reported to influence key mediators such as caspases and the PI3K/Akt pathway contributing to its investigation in cancer biology and neuroprotection The compound is soluble in water methanol and ethanol supporting its use in a variety of in vitro and in vivo research settings Its stability is maintained under sealed cool and dry storage conditions
Catalog No. 50-000-94370