Learn More
Medchemexpress LLC Ginsenoside F2 | 62025-49-4 | MFCD06410948 | 785.01 g/mol | C42H72O13 | 10MG
SDP
Catalog No. 5000237972
Encompass_Preferred
Ginsenoside F2 is a dammarane-type triterpenoid saponin and a metabolite of ginsenoside Rb1. It has been reported to induce apoptosis accompanied by protective autophagy in breast cancer stem cells and is used in biochemical and pharmacological research into apoptosis and autophagy mechanisms.
  • Natural dammarane-type ginsenoside, molecular formula C42H72O13.
  • Reported to induce apoptosis with protective autophagy in cancer stem cells.
  • Suitable for in vitro biochemical and pharmacological assays.
  • Molecular weight approximately 785.01 g/mol.
Catalog No. 50-002-37972 Supplier Medchemexpress LLC Supplier No. HY12584810MG
Ginsenoside F2 is a dammarane-type triterpenoid saponin and a metabolite of ginsenoside Rb1. It has been reported to induce apoptosis accompanied by protective autophagy in breast cancer stem cells and is used in biochemical and pharmacological research into apoptosis and autophagy mechanisms.
  • Natural dammarane-type ginsenoside, molecular formula C42H72O13.
  • Reported to induce apoptosis with protective autophagy in cancer stem cells.
  • Suitable for in vitro biochemical and pharmacological assays.
  • Molecular weight approximately 785.01 g/mol.