Learn More
Medchemexpress LLC Ginsenoside F2 | 62025-49-4 | MFCD06410948 | 100.0% | 785.01 g/mol | C42H72O13 | 1MG
SDP
Catalog No. 5000235578
Encompass_Preferred
Ginsenoside F2 is a dammarane-type ginsenoside and a metabolite of ginsenoside Rb1 reported to induce apoptosis accompanied by protective autophagy in breast cancer stem cells. Supplied as a research-grade powder analytical standard, it is suitable for in vitro biochemical studies and can be formulated for in vivo dosing using recommended vehicle systems.
Catalog No. 50-002-35578 Supplier Medchemexpress LLC Supplier No. HY1258481MG
Ginsenoside F2 is a dammarane-type ginsenoside and a metabolite of ginsenoside Rb1 reported to induce apoptosis accompanied by protective autophagy in breast cancer stem cells. Supplied as a research-grade powder analytical standard, it is suitable for in vitro biochemical studies and can be formulated for in vivo dosing using recommended vehicle systems.