Learn More
Medchemexpress LLC Ginsenoside F2 | 62025-49-4 | MFCD06410948 | 100.0% | 785.01 g/mol | C42H72O13 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12584810MM 1ML
Encompass_Preferred
Ginsenoside F2 is a dammarane-type ginsenoside and a metabolite of ginsenoside Rb1 that has been reported to induce apoptosis accompanied by protective autophagy in breast cancer stem cells. Supplied as an analytical standard for research use, the compound has a molecular formula of C42H72O13 and a molecular weight of approximately 785.01 g/mol, and is provided at high purity for analytical and biological studies.
  • High purity analytical standard (purity 99.95%).
  • Suitable for in vitro stock preparation at common concentrations (1, 5, 10 mM).
  • Reported solubility ≥2.5 mg/mL in selected in vivo vehicle systems.
  • Molecular formula C42H72O13 and molecular weight 785.01 g/mol.
  • Intended for research and analytical applications only.
Catalog No. 50-002-36561