Learn More
Cayman Chemical Ginsenoside F3 10MG
SDP
Catalog No. 502991918
Encompass_Preferred
A ginsenoside with immunomodulatory activity; increases proliferation of isolated mouse spleen cells and the production of IL-2 and IFN-γ, as well as decreases the production of IL-4 and IL-10, at 0.1-100 µM; increases concanavalin A-induced DNA binding activity of NF-κB in isolated mouse spleen cells at 10 µM
Catalog No. 50-299-1918 Supplier Cayman Chemical Supplier No. 3140910 MG
A ginsenoside with immunomodulatory activity; increases proliferation of isolated mouse spleen cells and the production of IL-2 and IFN-γ, as well as decreases the production of IL-4 and IL-10, at 0.1-100 µM; increases concanavalin A-induced DNA binding activity of NF-κB in isolated mouse spleen cells at 10 µM