Learn More
Medchemexpress LLC Ginsenoside F3 | 62025-50-7 | MFCD06410949 | 770.99 | 5 MG
SDP
Catalog No. 5000389084
Encompass_Preferred
Ginsenoside F3, a component of PPTGs (a minor saponin found in the leaves of Panax ginseng), demonstrates immunoenhancing activity. This is achieved by regulating the production and gene expression of both type 1 cytokines (IL-2, IFN-gamma) and type 2 cytokines (IL-4 and IL-10).
  • Immunoenhancing activity.
  • Regulates production and gene expression of type 1 cytokines (IL-2, IFN-gamma).
  • Regulates production and gene expression of type 2 cytokines (IL-4 and IL-10).
Catalog No. 50-003-89084 Supplier Medchemexpress LLC Supplier No. HYN06005MG
Ginsenoside F3, a component of PPTGs (a minor saponin found in the leaves of Panax ginseng), demonstrates immunoenhancing activity. This is achieved by regulating the production and gene expression of both type 1 cytokines (IL-2, IFN-gamma) and type 2 cytokines (IL-4 and IL-10).
  • Immunoenhancing activity.
  • Regulates production and gene expression of type 1 cytokines (IL-2, IFN-gamma).
  • Regulates production and gene expression of type 2 cytokines (IL-4 and IL-10).