Learn More
Medchemexpress LLC Ginsenoside F4 | 181225-33-2 | 5 MG
SDP
Catalog No. 5000418746
Encompass_Preferred
Ginsenoside F4 (GF4) is a ginseng saponin isolated from notoginseng or red ginseng. It demonstrates an inhibitory effect on human lymphocytoma JK cells by inducing apoptosis. GF4 also inhibits matrix metalloproteinase 13 (MMP 13) expression in IL-1β-treated chondrocytes and blocks cartilage breakdown in rabbit cartilage tissue culture, suggesting its therapeutic potential for preventing cartilage collagen matrix breakdown in diseased tissues.
  • Isolated from notoginseng or red ginseng
  • Induces apoptosis in human lymphocytoma JK cells
  • Inhibits matrix metalloproteinase 13 (MMP 13) expression
  • Blocks cartilage breakdown in rabbit cartilage tissue culture
  • Shows therapeutic potential for preventing cartilage collagen matrix breakdown
Catalog No. 50-004-18746 Supplier Medchemexpress LLC Supplier No. HYN25035MG
Ginsenoside F4 (GF4) is a ginseng saponin isolated from notoginseng or red ginseng. It demonstrates an inhibitory effect on human lymphocytoma JK cells by inducing apoptosis. GF4 also inhibits matrix metalloproteinase 13 (MMP 13) expression in IL-1β-treated chondrocytes and blocks cartilage breakdown in rabbit cartilage tissue culture, suggesting its therapeutic potential for preventing cartilage collagen matrix breakdown in diseased tissues.
  • Isolated from notoginseng or red ginseng
  • Induces apoptosis in human lymphocytoma JK cells
  • Inhibits matrix metalloproteinase 13 (MMP 13) expression
  • Blocks cartilage breakdown in rabbit cartilage tissue culture
  • Shows therapeutic potential for preventing cartilage collagen matrix breakdown