Learn More
Medchemexpress LLC Ginsenoside F5 | 189513-26-6 | MFCD22124710 | 99.9% | 770.99 | C41H70O13 | 5 MG
SDP
Catalog No. 5000242608
Encompass_Preferred
Ginsenoside F5 is a triterpenoid saponin isolated from flower buds of Panax ginseng. Reported studies show it inhibits HL-60 cell growth by inducing apoptosis. It is provided as a high-purity research reagent intended for in vitro biochemical and cellular assays.
  • High purity of 99.9%.
  • Induces apoptosis in HL-60 cells in reported studies.
  • Suitable for in vitro biochemical and cellular assays.
  • Molecular formula C41H70O13 and molecular weight 770.99.
  • Available in small milligram quantities for research use.
Catalog No. 50-002-42608 Supplier Medchemexpress LLC Supplier No. HY1082775MG
Ginsenoside F5 is a triterpenoid saponin isolated from flower buds of Panax ginseng. Reported studies show it inhibits HL-60 cell growth by inducing apoptosis. It is provided as a high-purity research reagent intended for in vitro biochemical and cellular assays.
  • High purity of 99.9%.
  • Induces apoptosis in HL-60 cells in reported studies.
  • Suitable for in vitro biochemical and cellular assays.
  • Molecular formula C41H70O13 and molecular weight 770.99.
  • Available in small milligram quantities for research use.