Learn More
Medchemexpress LLC HY-N0039 5mg Medchemexpress, Ginsenoside Rb1 CAS:41753-43-9 Purity:>98%
SDP
Catalog No. 502031295
Encompass_Preferred
Medchemexpress, HY-N0039 5mg Ginsenoside Rb1 CAS:41753-43-9 Ginsenoside Rb1, a main constituent of the root of Panax ginseng, inhibits Na+, K+-ATPase activity with an IC50 of 6.3±1.0 μM. Ginsenoside also inhibits IRAK-1 activation and phosphorylation of NF-κB p65 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1295 Supplier Medchemexpress LLC Supplier No. HYN00395MG
Medchemexpress, HY-N0039 5mg Ginsenoside Rb1 CAS:41753-43-9 Ginsenoside Rb1, a main constituent of the root of Panax ginseng, inhibits Na+, K+-ATPase activity with an IC50 of 6.3±1.0 μM. Ginsenoside also inhibits IRAK-1 activation and phosphorylation of NF-κB p65 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.