Learn More
Selleck Chemical LLC Ginsenoside Rb1 5mg 41753-43-9 Gypenoside
SDP
Supplier:  Selleck Chemical LLC S39245MG
Encompass_Preferred
Ginsenoside Rb1 (Gypenoside ?) is a protopanaxadiol that has diverse in vitro and in vivo effects, including neuroprotective, anti-inflammatory, and anti-obesity actions. Ginsenoside Rb1, a main constituent of the root of Panax ginseng, inhibits Na+, K+-ATPase activity with IC50 of 6.3¿1.0 ?M. Ginsenoside also inhibits IRAK-1 activation and phosphorylation of NF-?B p65. Ginsenoside Rb1 reduces the expressions of TLR3, TLR4 and TRAF-6, and down-regulates the levels of TNF-?, IFN-? and iNOS. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8349