Learn More
Cayman Chemical Ginsenoside Rb2, Purity: ≥95%, CAS: 11021-13-9, 10 mg
SDP
Catalog No. 502987807
Encompass_Preferred
A natural steroid glycoside with diverse effects; inhibits LPS-stimulated increases in VCAM-1 and ICAM-1 expression; reverses LPS-stimulated decreases in the expression of IκBα in HUVECs in a dose-dependent manner; reduces UVB-induced cytotoxicity and apoptotic nuclear fragmentation in HaCaT cells; lowers total cholesterol and triacylglycerol in 3T3-L1 adipocytes cultured under high fatty acid conditions to levels comparable to lovastatin
Catalog No. 50-298-7807 Supplier Cayman Chemical Supplier No. 2226510 MG
A natural steroid glycoside with diverse effects; inhibits LPS-stimulated increases in VCAM-1 and ICAM-1 expression; reverses LPS-stimulated decreases in the expression of IκBα in HUVECs in a dose-dependent manner; reduces UVB-induced cytotoxicity and apoptotic nuclear fragmentation in HaCaT cells; lowers total cholesterol and triacylglycerol in 3T3-L1 adipocytes cultured under high fatty acid conditions to levels comparable to lovastatin