Learn More
Cayman Chemical Ginsenoside Rb3, Purity: ≥98%, CAS: 68406-26-8, 100 mg
SDP
Catalog No. 502941856
Encompass_Preferred
A steroid glycoside with diverse biological activities; inhibits OGD-induced JNK-mediated activation of NF-κB and apoptosis in H9c2 mouse cardiac myoblasts; reduces myocardial apoptosis and production of ROS in a rat model of myocardial ischemia-reperfusion injury at 20 mg/kg; prevents increases in serum creatinine, BUN, MDA, and decreases in SOD activity and GSH levels, as well as inhibits renal cell apoptosis in a mouse model of cisplatin-induced nephrotoxicity at 10 and 20 mg/kg; decreases the abundance of cancer cachexia-associated gut microbiota and the number of polyps in the ApcMin/+ mouse model of colon cancer; decreases immobility time in the forced swim and tail suspension tests in mice
Catalog No. 50-294-1856 Supplier Cayman Chemical Supplier No. 29005100 MG
A steroid glycoside with diverse biological activities; inhibits OGD-induced JNK-mediated activation of NF-κB and apoptosis in H9c2 mouse cardiac myoblasts; reduces myocardial apoptosis and production of ROS in a rat model of myocardial ischemia-reperfusion injury at 20 mg/kg; prevents increases in serum creatinine, BUN, MDA, and decreases in SOD activity and GSH levels, as well as inhibits renal cell apoptosis in a mouse model of cisplatin-induced nephrotoxicity at 10 and 20 mg/kg; decreases the abundance of cancer cachexia-associated gut microbiota and the number of polyps in the ApcMin/+ mouse model of colon cancer; decreases immobility time in the forced swim and tail suspension tests in mice