Learn More
TARGETMOL CHEMICALS INC GINSENOSIDE RB3 50MG
SDP
Catalog No. 502576859
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. Ginsenoside Rb3 (Gypenoside IV) is a natural triterpenoid saponin exhibiting inhibition effect on TNF(alpha)-induced NF-(kappa)B transcriptional activity. It also inhibits the induction of COX-2 and iNOS mRNA. purity: 99%
Catalog No. 50-257-6859 Supplier TARGETMOL CHEMICALS INC Supplier No. T391850MG
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. Ginsenoside Rb3 (Gypenoside IV) is a natural triterpenoid saponin exhibiting inhibition effect on TNF(alpha)-induced NF-(kappa)B transcriptional activity. It also inhibits the induction of COX-2 and iNOS mRNA. purity: 99%