Learn More
Medchemexpress LLC HY-N0041 5mg Medchemexpress, Ginsenoside Rb3 CAS:68406-26-8 Purity:>98%
SDP
Catalog No. 501933366
Encompass_Preferred
Medchemexpress, HY-N0041 5mg Ginsenoside Rb3 CAS:68406-26-8 Ginsenoside Rb3 is extracted from steamed Panax notoginseng. Ginsenoside Rb3 exhibits inhibitory effect on TNFα-induced NF-κB transcriptional activity with an IC 50 of 8.2 μM in 293T cell lines. Ginsenoside Rb3 also inhibits the induction of COX-2 and iNOS mRNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3366 Supplier Medchemexpress LLC Supplier No. HYN00415MG
Medchemexpress, HY-N0041 5mg Ginsenoside Rb3 CAS:68406-26-8 Ginsenoside Rb3 is extracted from steamed Panax notoginseng. Ginsenoside Rb3 exhibits inhibitory effect on TNFα-induced NF-κB transcriptional activity with an IC 50 of 8.2 μM in 293T cell lines. Ginsenoside Rb3 also inhibits the induction of COX-2 and iNOS mRNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.