Learn More
Medchemexpress LLC HY-N0042 5mg Medchemexpress, Ginsenoside Rc CAS:11021-14-0 Purity:>98%
SDP
Catalog No. 501933367
Encompass_Preferred
Medchemexpress, HY-N0042 5mg Ginsenoside Rc CAS:11021-14-0 Ginsenoside Rc, one of major Ginsenosides from Panax ginseng, enhances GABA receptor A ( GABA A )-mediated ion channel currents (I GABA ). Ginsenoside Rc inhibits the expression of TNF-α and IL-1β . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3367 Supplier Medchemexpress LLC Supplier No. HYN00425MG
Medchemexpress, HY-N0042 5mg Ginsenoside Rc CAS:11021-14-0 Ginsenoside Rc, one of major Ginsenosides from Panax ginseng, enhances GABA receptor A ( GABA A )-mediated ion channel currents (I GABA ). Ginsenoside Rc inhibits the expression of TNF-α and IL-1β . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.