Learn More
Cayman Chemical GInsenosIde Rd 10mg
SDP
Catalog No. 502969447
Encompass_Preferred
A protopanaxadiol that has diverse in vitro and in vivo effects, including cardioprotective, neuroprotective, and anti-inflammatory actions
Catalog No. 50-296-9447 Supplier Cayman Chemical Supplier No. 1532910 MG
A protopanaxadiol that has diverse in vitro and in vivo effects, including cardioprotective, neuroprotective, and anti-inflammatory actions