Learn More
Medchemexpress LLC HY-N0043 5mg Medchemexpress, Ginsenoside Rd CAS:52705-93-8 Purity:>98%
SDP
Catalog No. 501933368
Encompass_Preferred
Medchemexpress, HY-N0043 5mg Ginsenoside Rd CAS:52705-93-8 Ginsenoside Rd inhibits TNFα-induced NF-κB transcriptional activity with an IC 50 of 12.05±0.82 μM in HepG2 cells. Ginsenoside Rd inhibits expression of COX-2 and iNOS mRNA. Ginsenoside Rd also inhibits Ca 2+ influx. Ginsenoside Rd inhibits CYP2D6 , CYP1A2 , CYP3A4 , and CYP2C9 , with IC 50 s of 58.0±4.5 μM, 78.4±5.3 μM, 81.7±2.6 μM, and 85.1±9.1 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3368 Supplier Medchemexpress LLC Supplier No. HYN00435MG
Medchemexpress, HY-N0043 5mg Ginsenoside Rd CAS:52705-93-8 Ginsenoside Rd inhibits TNFα-induced NF-κB transcriptional activity with an IC 50 of 12.05±0.82 μM in HepG2 cells. Ginsenoside Rd inhibits expression of COX-2 and iNOS mRNA. Ginsenoside Rd also inhibits Ca 2+ influx. Ginsenoside Rd inhibits CYP2D6 , CYP1A2 , CYP3A4 , and CYP2C9 , with IC 50 s of 58.0±4.5 μM, 78.4±5.3 μM, 81.7±2.6 μM, and 85.1±9.1 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.