Learn More
Cayman Chemical Ginsenoside Re, Purity: ≥95%, CAS: 52286-59-6, 10 mg
SDP
Catalog No. 502972301
Encompass_Preferred
A panaxatriol saponin that has diverse in vitro and in vivo effects, including vasorelaxant, antioxidant, antihyperlipidemic, and angiogenic actions; improves the effectiveness of chemotherapeutic agents by decreasing the expression of Multidrug Resistance Protein 1 and inhibiting P-glycoprotein
Catalog No. 50-297-2301 Supplier Cayman Chemical Supplier No. 1533010 MG
A panaxatriol saponin that has diverse in vitro and in vivo effects, including vasorelaxant, antioxidant, antihyperlipidemic, and angiogenic actions; improves the effectiveness of chemotherapeutic agents by decreasing the expression of Multidrug Resistance Protein 1 and inhibiting P-glycoprotein