Learn More
Medchemexpress LLC HY-N0044 5mg Medchemexpress, Ginsenoside Re CAS:52286-59-6 Purity:>98%
SDP
Catalog No. 501933369
Encompass_Preferred
Medchemexpress, HY-N0044 5mg Ginsenoside Re CAS:52286-59-6 Ginsenoside Re (Ginsenoside B2) is an extract from Panax notoginseng. Ginsenoside Re decreases the β-amyloid protein ( Aβ ). Ginsenoside Re plays a role in antiinflammation through inhibition of JNK and NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3369 Supplier Medchemexpress LLC Supplier No. HYN00445MG
Medchemexpress, HY-N0044 5mg Ginsenoside Re CAS:52286-59-6 Ginsenoside Re (Ginsenoside B2) is an extract from Panax notoginseng. Ginsenoside Re decreases the β-amyloid protein ( Aβ ). Ginsenoside Re plays a role in antiinflammation through inhibition of JNK and NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.