Learn More
Cayman Chemical Ginsenoside Rf, Purity: ≥95%, CAS: 52286-58-5, 1 mg
SDP
Catalog No. 502994649
Encompass_Preferred
A natural steroid glycoside with diverse effects; inhibits production of IL-1β, IL-6, TNF-α, NO, and ROS in HT-29 and RAW 264.7 cells in a dose-dependent manner; inhibits N-type calcium channels in rat sensory neurons (IC50 = 40 μM); has antinociceptive effects in mice in the acetic acid abdominal constriction test and in the biphasic formalin test (ED50s = 56 and 129 mg/kg, respectively); reduces ear swelling by 34.8% in a mouse model of contact dermatitis
Catalog No. 50-299-4649 Supplier Cayman Chemical Supplier No. 236671 MG
A natural steroid glycoside with diverse effects; inhibits production of IL-1β, IL-6, TNF-α, NO, and ROS in HT-29 and RAW 264.7 cells in a dose-dependent manner; inhibits N-type calcium channels in rat sensory neurons (IC50 = 40 μM); has antinociceptive effects in mice in the acetic acid abdominal constriction test and in the biphasic formalin test (ED50s = 56 and 129 mg/kg, respectively); reduces ear swelling by 34.8% in a mouse model of contact dermatitis