Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ginsenoside Rf | 52286-58-5 | 801.01 | 50 MG
SDP

Catalog No. 5000388430
Encompass_Preferred

Ginsenoside Rf is a trace component of ginseng root. It is a saponin that rapidly and reversibly inhibits N-type and other high-threshold Ca2+ channels in rat sensory neurons. It is suitable for various laboratory applications.

  • Inhibits N-type Ca2+ channels
  • Trace component of ginseng root
  • Effect largely eliminated by pretreatment with pertussis toxin
  • Inhibits Ca2+ channels in hybrid F-11 cell line
  • Appears as a white to off-white solid
  • Target: N-type calcium channel
  • Produces dose-dependent antinociception in mice
  • Soluble in DMSO and ethanol for in vitro applications

Catalog No. 50-003-88430 Supplier Medchemexpress LLC Supplier No. HYN060150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ginsenoside Rf is a trace component of ginseng root. It is a saponin that rapidly and reversibly inhibits N-type and other high-threshold Ca2+ channels in rat sensory neurons. It is suitable for various laboratory applications.

  • Inhibits N-type Ca2+ channels
  • Trace component of ginseng root
  • Effect largely eliminated by pretreatment with pertussis toxin
  • Inhibits Ca2+ channels in hybrid F-11 cell line
  • Appears as a white to off-white solid
  • Target: N-type calcium channel
  • Produces dose-dependent antinociception in mice
  • Soluble in DMSO and ethanol for in vitro applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.