Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginsenoside Rf, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014440 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4440 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4440 Supplier MilliporeSigma Supelco Supplier No. 658395MG
Add to Cart
Add to Cart
Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 52286-58-5
Physical Form Neat
Linear Formula C42H72O14
MDL Number MFCD00210509
Synonym (3 beta,6 alpha,12 beta)-3,12,20-Trihydroxydammar-24-en-6-yl 2-O-beta-D-glucopyranosyl-beta-D-glucopyranoside; Panaxoside Rf
Type Analytical Standard
Quantity 5 mg
Formula Weight 801.01
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.