Learn More
Cayman Chemical Ginsenoside Rg2, Purity: ≥98%, CAS: 52286-74-5, 10 mg
SDP
Catalog No. 502972354
Encompass_Preferred
A protopanaxatriol that has diverse in vitro and in vivo effects, including neuroprotective, anti-inflammatory, and anti-diabetic actions; protects against DNA damage and apoptosis induced by ultraviolet light
Catalog No. 50-297-2354 Supplier Cayman Chemical Supplier No. 1533110 MG
A protopanaxatriol that has diverse in vitro and in vivo effects, including neuroprotective, anti-inflammatory, and anti-diabetic actions; protects against DNA damage and apoptosis induced by ultraviolet light