Learn More
Medchemexpress LLC HY-N0602 5mg Medchemexpress, Ginsenoside Rg2 CAS:52286-74-5 Purity:>98%
SDP
Catalog No. 501933634
Encompass_Preferred
Medchemexpress, HY-N0602 5mg Ginsenoside Rg2 CAS:52286-74-5 Ginsenoside Rg2 is one of the major active components of ginseng. Ginsenoside Rg2 acts as a NF-κB inhibitor. Ginsenoside Rg2 also reduces Aβ 1-42 accumulation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3634 Supplier Medchemexpress LLC Supplier No. HYN06025MG
Medchemexpress, HY-N0602 5mg Ginsenoside Rg2 CAS:52286-74-5 Ginsenoside Rg2 is one of the major active components of ginseng. Ginsenoside Rg2 acts as a NF-κB inhibitor. Ginsenoside Rg2 also reduces Aβ 1-42 accumulation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.