Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginsenoside Rg2, ≥94% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014442 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4442 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4442 Supplier MilliporeSigma Supelco Supplier No. 0817110MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ginsenoside Rg2 is a saponin and a protopanaxatriol-type compound found in the root and stem leaves of ginseng.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 52286-74-5
Linear Formula C42H72O13
MDL Number MFCD00210511
Synonym (3 beta,6 alpha,12 beta)-3,12,20-Trihydroxydammar-24-en-6-yl 2-O-(6-deoxy-alpha-L-mannopyranosyl)-beta-D-glucopyranoside; Chikusetsusaponin I; Panaxoside Rg2; Prosapogenin C2
Type Analytical Standard
Quantity 10 mg
Formula Weight 785.01
Percent Purity ≥94% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.