Learn More
Lkt Laboratories Ginsenoside Rg3, ≥98%, 5MG
SDP
Supplier:  Lkt Laboratories G35565MG
Encompass_Preferred
Triterpene saponin found in species of Panax; γ2 GABA-A agonist, Kv7.1 K+ channel activator, α10 nAChR antagonist. CAS: 38243-03-7. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4917