Learn More
Medchemexpress LLC Ginsenoside Rg4 | 126223-28-7 | 5 MG
SDP
Catalog No. 5000420207
Encompass_Preferred
Ginsenoside Rg4 is an orally active protopanaxatriol type ginsenoside. It can activate PI3K, AKT, and GSK-3β signaling, and inhibit ROS and inflammatory cytokine levels. It is used in research for inflammation, infection, and metabolic diseases like sepsis and lung inflammation.
  • Activates PI3K, AKT, and GSK-3β signaling
  • Inhibits ROS and inflammatory cytokine levels
  • Used for research on inflammation, infection, and metabolic diseases
  • Decreases expressions of toll-like receptor (TLR) 4 and TNF-α levels
  • Increases cell viability via PI3K/AKT signaling activation
  • Promotes hair-inductive properties of DP cells
  • Inhibits inflammation and prolongs survival in sepsis models
  • Attenuates inflammation in pulmonary inflammation models
Catalog No. 50-004-20207 Supplier Medchemexpress LLC Supplier No. HYN65805MG
Ginsenoside Rg4 is an orally active protopanaxatriol type ginsenoside. It can activate PI3K, AKT, and GSK-3β signaling, and inhibit ROS and inflammatory cytokine levels. It is used in research for inflammation, infection, and metabolic diseases like sepsis and lung inflammation.
  • Activates PI3K, AKT, and GSK-3β signaling
  • Inhibits ROS and inflammatory cytokine levels
  • Used for research on inflammation, infection, and metabolic diseases
  • Decreases expressions of toll-like receptor (TLR) 4 and TNF-α levels
  • Increases cell viability via PI3K/AKT signaling activation
  • Promotes hair-inductive properties of DP cells
  • Inhibits inflammation and prolongs survival in sepsis models
  • Attenuates inflammation in pulmonary inflammation models