Learn More
Medchemexpress LLC HY-N0908 5mg Medchemexpress, Ginsenoside Rg5 CAS:186763-78-0 Purity:>98%
SDP
Catalog No. 501933791
Encompass_Preferred
Medchemexpress, HY-N0908 5mg Ginsenoside Rg5 CAS:186763-78-0 Ginsenoside Rg5 is the main component of Red ginseng. Ginsenoside blocks binding of IGF-1 to its receptor with an IC 50 of ~90 nM. Ginsenoside Rg5 also inhibits the mRNA expression of COX-2 via suppression of the DNA binding activities of NF-κB p65 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3791 Supplier Medchemexpress LLC Supplier No. HYN09085MG
Medchemexpress, HY-N0908 5mg Ginsenoside Rg5 CAS:186763-78-0 Ginsenoside Rg5 is the main component of Red ginseng. Ginsenoside blocks binding of IGF-1 to its receptor with an IC 50 of ~90 nM. Ginsenoside Rg5 also inhibits the mRNA expression of COX-2 via suppression of the DNA binding activities of NF-κB p65 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.