Learn More
Selleck Chemical LLC Ginsenoside Rg5 E2459-5MG
SDP
Catalog No. 5000040016
Encompass_Preferred
Ginsenoside Rg5 the main component of Red ginseng blocks binding of IGF-1 to its receptor with an IC50 of 90 nM Ginsenoside Rg5 also inhibits the mRNA expression of COX-2 via suppression of the DNA binding activities of NF- B p65
Catalog No. 50-000-40016 Supplier Selleck Chemical LLC Supplier No. E24595MG
Ginsenoside Rg5 the main component of Red ginseng blocks binding of IGF-1 to its receptor with an IC50 of 90 nM Ginsenoside Rg5 also inhibits the mRNA expression of COX-2 via suppression of the DNA binding activities of NF- B p65