Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginsenoside Rg5, ≥90.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013395 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3395 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3395 Supplier MilliporeSigma Supelco Supplier No. 430165MG
Add to Cart
Add to Cart

Ginsenoside Rg5 is a diol-containing ginsenoside, that occurs naturally in Panax ginseng. It is found to be a promising agent in the treatment of Alzheimer′s disease.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 186763-78-0
Linear Formula C42H70O12
Synonym (3 beta, 12 beta,20 E)-12-Hydroxydammara-20(22),24-dien-3-yl 2-O-beta-D-glucopyranosyl-beta-D-glucopyranoside
Type Analytical Standard
Quantity 5 mg
Formula Weight 767.00
Percent Purity ≥90.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.