Learn More
Medchemexpress LLC HY-N0907 1mg Medchemexpress, Ginsenoside Rg6 CAS:147419-93-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN09071MG
Encompass_Preferred
Medchemexpress, HY-N0907 1mg Ginsenoside Rg6 CAS:147419-93-0 Ginsenoside Rg6 is the component isolated from notoginseng. Ginsenoside Rg6 inhibits TNF-α-induced NF-κB transcriptional activity with an IC50 of 29.34±2.22 μM in HepG2 cells. Ginsenoside Rg6 also exhibits apoptosis-inducing effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2828