Learn More
Medchemexpress LLC HY-N0604 5mg Medchemexpress, Ginsenoside Rh1 CAS:63223-86-9 Purity:>98%
SDP
Catalog No. 501933635
Encompass_Preferred
Medchemexpress, HY-N0604 5mg Ginsenoside Rh1 CAS:63223-86-9 Ginsenoside Rh1 (Prosapogenin A2; Sanchinoside B2; Sanchinoside Rh1) is isolated from the root of Panax Ginseng. Ginsenoside Rh1 inhibits the expression of PPAR-γ , TNF-α , IL-6 , and IL-1β . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3635 Supplier Medchemexpress LLC Supplier No. HYN06045MG
Medchemexpress, HY-N0604 5mg Ginsenoside Rh1 CAS:63223-86-9 Ginsenoside Rh1 (Prosapogenin A2; Sanchinoside B2; Sanchinoside Rh1) is isolated from the root of Panax Ginseng. Ginsenoside Rh1 inhibits the expression of PPAR-γ , TNF-α , IL-6 , and IL-1β . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.