Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ginsenoside Rh2 (20(S)-ginsenoside Rh2) | 78214-33-2 | 622.87 | 100 MG
SDP

Catalog No. 5000389712
Encompass_Preferred

Ginsenoside Rh2 (20(S)-Ginsenoside Rh2) induces the activation of caspase-8 and caspase-9. It induces cancer cell apoptosis in a multi-path manner, making it a promising candidate for anti-tumor drug development. Ginsenoside Rh2 triggers p53-dependent Fas expression and consequent activation of caspase-8 and p53-independent caspase-9-mediated intrinsic pathway to cause cancer cell death.

  • Purity: 99.28%
  • Appearance: Solid
  • Color: White to off-white
  • Formula: C36H62O8
  • Structure classification: Terpenoids, triterpenes
  • Initial source: Plants, Araliaceae, Panax ginseng C. A. Meyer
  • Endogenous metabolite: Human gut microbiota metabolites
  • Solubility (in vitro): DMSO: 50 mg/mL (80.27 mM; requires ultrasonic; use newly opened DMSO)
  • Usage: For research use only

Catalog No. 50-003-89712 Supplier Medchemexpress LLC Supplier No. HYN0605100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ginsenoside Rh2 (20(S)-Ginsenoside Rh2) induces the activation of caspase-8 and caspase-9. It induces cancer cell apoptosis in a multi-path manner, making it a promising candidate for anti-tumor drug development. Ginsenoside Rh2 triggers p53-dependent Fas expression and consequent activation of caspase-8 and p53-independent caspase-9-mediated intrinsic pathway to cause cancer cell death.

  • Purity: 99.28%
  • Appearance: Solid
  • Color: White to off-white
  • Formula: C36H62O8
  • Structure classification: Terpenoids, triterpenes
  • Initial source: Plants, Araliaceae, Panax ginseng C. A. Meyer
  • Endogenous metabolite: Human gut microbiota metabolites
  • Solubility (in vitro): DMSO: 50 mg/mL (80.27 mM; requires ultrasonic; use newly opened DMSO)
  • Usage: For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.