Learn More
Medchemexpress LLC HY-N0605 5mg Medchemexpress, Ginsenoside Rh2 CAS:78214-33-2 Purity:>98%
SDP
Catalog No. 501933636
Encompass_Preferred
Medchemexpress, HY-N0605 5mg Ginsenoside Rh2 CAS:78214-33-2 Ginsenoside Rh2 is isolated from the root of Ginseng. Ginsenoside Rh2 induces the activation of caspase-8 and caspase-9 . Ginsenoside Rh2 induces cancer cell apoptosis in a multi-path manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3636 Supplier Medchemexpress LLC Supplier No. HYN06055MG
Medchemexpress, HY-N0605 5mg Ginsenoside Rh2 CAS:78214-33-2 Ginsenoside Rh2 is isolated from the root of Ginseng. Ginsenoside Rh2 induces the activation of caspase-8 and caspase-9 . Ginsenoside Rh2 induces cancer cell apoptosis in a multi-path manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.