Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginsenoside Rh2, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015541 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5541 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5541 Supplier MilliporeSigma Supelco Supplier No. 7365810MG
Add to Cart
Add to Cart

Ginsenoside Rh2 is an active component of Red P. ginseng. It is reported to be obtained as a metabolite of ginsenoside Rg3, when anaerobically incubated with human fecal microflora.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 78214-33-2
Physical Form Neat
Linear Formula C36H62O8
MDL Number MFCD00800712
Synonym (3 beta,12 beta)-12,20-Dihydroxydammar-24-en-3-yl-beta-D-glucopyranoside; 20-Ginsenoside-Rh2
Type Analytical Standard
Quantity 10 mg
Formula Weight 622.87
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.