Learn More
Medchemexpress LLC HY-N0606 5mg Medchemexpress, Ginsenoside Rh3 CAS:105558-26-7 Purity:>98%
SDP
Catalog No. 501933637
Encompass_Preferred
Medchemexpress, HY-N0606 5mg Ginsenoside Rh3 CAS:105558-26-7 Ginsenoside Rh3 is a bacterial metabolite of Ginsenoside Rg5. Ginsenoside Rh3 treatment in human retinal cells induces Nrf2 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3637 Supplier Medchemexpress LLC Supplier No. HYN06065MG
Medchemexpress, HY-N0606 5mg Ginsenoside Rh3 CAS:105558-26-7 Ginsenoside Rh3 is a bacterial metabolite of Ginsenoside Rg5. Ginsenoside Rh3 treatment in human retinal cells induces Nrf2 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.